Learn More
Ansell™ AlphaTec™ 5000 Ultrasonically Welded and Taped
AlphaTec™ 5000 Ultrasonically welded and taped made of highly visible innovative material is strong, durable and suitable for workers in extremely hazardous areas, including HAZMAT response teams.
£1125.00
Specifications
Product Type | Chemical Protective Clothing |
---|---|
Gender | Unisex |
Material | Multi-layer Barrier Laminate Fabric |
Color | Orange |
Disposable | Reusable |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17120905
|
Ansell™
OR50T-00198-02 |
Small |
£1125.00
Pack of 10 |
Estimated Shipment: 10-12-2025
Log in to see stock. |
|||||
17130905
|
Ansell™
OR50T-00198-03 |
Medium |
£1125.00
Pack of 10 |
Estimated Shipment: 10-12-2025
Log in to see stock. |
|||||
17140905
|
Ansell™
OR50T-00198-04 |
Large |
£1125.00
Pack of 10 |
Estimated Shipment: 10-12-2025
Log in to see stock. |
|||||
17150905
|
Ansell™
OR50T-00198-05 |
X-Large |
£1125.00
Pack of 10 |
Estimated Shipment: 10-12-2025
Log in to see stock. |
|||||
17160905
|
Ansell™
OR50T-00198-06 |
2X-Large |
£1125.00
Pack of 10 |
Estimated Shipment: 10-12-2025
Log in to see stock. |
|||||
17170905
|
Ansell™
OR50T-00198-07 |
3X-Large |
£1125.00
Pack of 10 |
Estimated Shipment: 10-12-2025
Log in to see stock. |
|||||
Description
Made of highly visible innovative material is strong, durable and suitable for workers in extremely hazardous areas, including HAZMAT response teams.
- Outstanding permeation performance to numerous organic and inorganic chemicals, biological hazards and chemical warfare agents
- Exceptional barrier protection, providing andgt;480 minutes breakthrough time against 14 of 15 chemicals listed in EN ISO 6529
- Highly visible bright orange colour for improved worker safety
- Welded and taped seams, our highest barrier to liquids and particulates
- Designed to protect, typical innovative features include dual zip systems
- Multi-layer barrier material which is lightweight, yet strong and durable
- Anti-static, tested according to EN 1149-5
Specifications
Chemical Protective Clothing | |
Multi-layer Barrier Laminate Fabric | |
Reusable | |
Double, Elastic | |
Standard | |
Welded, Taped | |
CE CATEGORY III | |
CE CATEGORY III (Type 3-B, Type 4-B, Type 5-B, EN 1073-2, EN 14126, EN 1149-5) | |
Contaminated Environments, Chemical Handling, Filling, blending and charging of raw materials, Transferring liquids and solids, Opening and draining pumps valves or lines, Loading and unloading of proces equipment, Raw material sample collection, Concentration mixing and palletisation, Hazmat response, Assembly and disassembly of large pipes, Unexpected leakages spills or other releases |
Unisex | |
Orange | |
Elastic | |
Collared | |
Double Zipper | |
Non-sterile | |
Chemicals, Anti-static, Particle | |
None |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.