missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 38001PP Medium Duty Chemical-resistant Gloves with Gauntlet Cuff

AlphaTec™ 38001PP Medium Duty Chemical-resistant Gloves with Gauntlet Cuff are made of water-based butyl and boast a raised diamond grip for precise industrial applications.

Brand:  Ansell 38001PP110

48.83 GBP valid until 2025-09-30
Use promo code "24297" to get your promotional price.


 View more versions of this product

Product Code. 18576574

  • £65.85 / 1 pair
Only null left
Estimated Shipment: 17-09-2025
Add to basket
This item is not returnable. View return policy

Description

Description

  • High flexibility and tactility performance vs. standard butyl
  • Raised diamond pattern for enhanced grip
  • High performance against highly aggressive chemicals including solvents, aldehydes, ketones, esters and concentrated mineral acids
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Industrial
Yes
11
0.35 mm
14 in.
Black
Butyl
14 mil
Gauntlet
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.