Miele Professional™ Additional Suction Lance
Suction lance for external canisters for PLW 8636
Additional Details:
Additional Details: Weight: 0.10000kg
Product Code. 30533217
Quantity Price
1 £ 599.0 / 1 sheet
Estimated Shipment
08-07-2025
Add to basket
Description and Specification
Specification
Product Type Filter
For Use With (Equipment) Washer-Disinfectors
Quantity 1 Ea.
  • Suction lance for external canisters for PLW 8636
  • For 10 - 20 L canisters, positioned outside the machine
  • The suction lance features level detection
  • Installed by Customer Service