SPEX Certiprep Acrolein, Deionized Water Matrix, SPEX CertiPrep™


Overview
Code: NEW

Additional Details:
Additional Details: CAS Number: 7732-18-5 Weight: 0.05000kg


Quantity 1 mL

Product Code. 15200064

Quantity Price
1 £ 89.65 / 1mL
Estimated Shipment
16-09-2025
Add to basket


Description and Specification

Specification

Concentration 1000 μg/mL
Solvent or Matrix Deionized Water
Quantity 1 mL
Safety and Handling

[Acrolein]

[Danger]

[{code=EYE DAM. 1, sequence=1, description=Serious eye damage/eye irritation Category 1}, {code=SKIN CORR. 1B, sequence=2, description=Skin corrosion/irritation Category 1 B}]

[{code=H314, sequence=1, description=Causes severe skin burns and eye damage.}, {code=H318, sequence=2, description=Causes serious eye damage.}]

[{code=P303+P361+P353, sequence=51, description=IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.}, {code=P305+P351+P338, sequence=52, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P310, sequence=53, description=Immediately call a POISON CENTER/doctor/.}, {code=P321, sequence=54, description=Specific treatment (see on this label).}, {code=P405, sequence=55, description=Store locked up.}, {code=P501b, sequence=56, description=Dispose of contents/container in accordance with local/regional/national/international regulations.}]

[{code=MIXTURE LIST, sequence=1, description=Contains : acrylaldehyde}]

[{code=GHS05, sequence=1, description=CORROSION}]


Product Identifier
  • Acrolein

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314- Causes severe skin burns and eye damage.
  • H318- Causes serious eye damage.

Precautionary Statement
  • P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310- Immediately call a POISON CENTER/doctor/.
  • P321- Specific treatment (see on this label).
  • P405- Store locked up.
  • P501b- Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental Information
  • MIXTURE LIST- Contains : acrylaldehyde