Greiner Bio-One 96-well Conical Bottom (V) Polypropylene Microplates

To significantly improve data quality and sensitivity in biochemical assays. Greiner Bio-One 96-Well Conical Bottom (V) Polypropylene Microplates is free of detectable DNase, RNase, human DNA.

Overview
Code: Z1

Additional Details:
Additional Details: Weight: 3.83000kg



Product Code. 13092143

 N/A
N/A

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description and Specification

Specification

Certifications/Compliance Free of detectable DNase/RNase, human DNA and pyrogens.
Format Standard
Lid Without Lid
Array 8 x 12
Autoclavable Not Autoclavable
Color Yellow
Material Polypropylene
No. of Wells 96
Sterility Non-sterile
Well Shape Conical Bottom (V-well)
Quantity 100/Cs.
Dimensions (L x W x H) 128 x 85 x 15 mm

  • Uniform external dimensions
  • Well to well spacing of 9mm
  • Features alphanumeric coding
  • Sealable with adhesive films and heat seal