missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One 96-well Conical Bottom (V) Polypropylene Microplates

To significantly improve data quality and sensitivity in biochemical assays. Greiner Bio-One 96-Well Conical Bottom (V) Polypropylene Microplates is free of detectable DNase, RNase, human DNA.
Brand: Greiner Bio-One 651209
This item is not returnable.
View return policy
Description
- Uniform external dimensions
- Well to well spacing of 9mm
- Features alphanumeric coding
- Sealable with adhesive films and heat seal

Specifications
Free of detectable DNase/RNase, human DNA and pyrogens. | |
Without Lid | |
Not Autoclavable | |
Polypropylene | |
50 to 335 μL | |
Conical Bottom (V-well) | |
100 | |
100/Cs. | |
Chimney |
Standard | |
8 x 12 | |
Black | |
96 | |
Non-sterile | |
4800 x g (swinging bucket rotor) | |
10 | |
-196°C to +121°C | |
128 x 85 x 15 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction