Learn More
Greiner Bio-One 96-well Chimney designed Round (U) Bottom Microplates
For high-throughput screening in cell based test systems.
Brand:  Greiner Bio-One 650207
 View more versions of this product
Product Code. 10433075
  • £142.30 / Pack of 100
Only null left
Estimated Shipment: 15-12-2025
Add to basket
Description
Description
  • Non-pyrogenic
  • Conical well design with flat bottom
  • Ensures accurate pipetting
  • Reduces dead volume of well
  • Free of detectable DNase, RNase, human DNA
  • Prevents wicking
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Free of detectable DNase/RNase, human DNA and pyrogens.
8 x 12
White
96
Non-sterile
Round Bottom (U-well)
100
100/Cs.
128 x 85 x 15 mm
Standard
Not Autoclavable
Polypropylene
50 to 300 μL
355 μL
4800 x g (swinging bucket rotor)
10
-196°C to +121°C