missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 384-Well, Flat-Bottom Microplate, Non-Binding Coated
GREENER_CHOICE

Color:White; Sterility:Non-sterile; Lid:Without Lid

Brand:  Corning™ 3767

1481.24 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15762907

  • £1491.61 / Pack of 100
Only null left
Estimated Shipment: 12-01-2026
Add to basket
This item is not returnable. View return policy

Description

Description

Suited for fluorescent and luminescent assays using either top or bottom detection microplate readers.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Standard
16 x 24
Polystyrene
Non-sterile
Flat Bottom (F-well)
25/Pk.
11.6 mm
Without Lid
White, Clear Bottom
384
Non-binding
100/Cs.
127.8 x 85.5 x 14.35 mm (5 x 3.36 x 0.56 in.)
Top/bottom: 3.63/2.82 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.