14.54 GBP valid until 2025-12-31
Use promo code "24302" to get your promotional price.
Use promo code "24302" to get your promotional price.
Certifications/Compliance (Europe) | CE 2797 PPE Category III according to Regulation (EU) 2016/425 EEC |
---|---|
Chemotherapeutic Rated | 14 Chemotherapy Drugs |
Chlorinated | No |
Accelerator Free | No |
Applications | Laboratory |
AQL | 1 |
Color | Dark Blue |
Disposable | Single-use |
Exterior Finish | Fingertip Textured |
Finger Thickness (English) | 6 mil |
Finger Thickness (Metric) | 0.15 mm |
Latex Free | Yes |
Material | Nitrile |
Palm Thickness (English) | 4.7 mil |
Palm Thickness (Metric) | 0.12 mm |
Powdered | Powder Free |
Product Type | Nitrile Examination Glove |
Size | X-Large |
Sterility | Non-sterile |
Thickness (English) | 6 mil |
Thickness (Metric) | 0.15 mm |
FDA 510 (k) | Yes |
Glove Cuff Style | Beaded |
Model | LFS121XL |
Series | LFS121 |
Tensile Strength | 42 MPa (Target) |
Length (English) | 12 in. |
Length (Metric) | 30.5 cm |