Use promo code "25339" to get your promotional price.
Thermo Scientific™ SureSTART™ 9 mm Screw Caps, Level 2 High-throughput Applications
Use Level 2 Thermo Scientific™ SureSTART™ 9 mm Screw Caps with screw vials that have a 9 mm opening for your high-throughput applications, when robustness and reproducibility are key.
20.22 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Use promo code "25339" to get your promotional price.
Additional Details:
Weight:
0.09000kg
Specification
| Color | Green |
|---|---|
| Material | Polypropylene |
| Pre-slit | No |
| Septum | White Silicone/Red PTFE |
| Hardness | 45 Shore |
| Product Line | SureSTART Level 2 |
| Quantity | 100 Pk. |
| Thickness (Metric) | 1 mm |
| Closure Size | 9 mm |
| For Use With (Equipment) | Screw Thread Vial |
| Green Features | ACT Ecolabel (Silver) |
| Certifications/Compliance | Specification Certified |
| Product Type | AVCS Screw Cap |