Memmert™ Support Basket with Perforated Mounting Shelf
Support basket with perforated mounting shelf for conical flasks clips
Overview
103.74 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Additional Details:
Additional Details: Weight: 0.01100kg
Product Code. 17120345
Quantity Price
1 £ 110.0 / Each
Estimated Shipment
15-09-2025
Add to basket
Description and Specification
Specification
For Use With (Equipment) WTB24
Type Support Basket With Perforated Mounting Shelf (Stainless Steel)
Material Stainless Steel
  • Use of test tube racks not possible
  • Has grid perforations to take clips for conical flasks