Thermo Scientific™ Nunc™ Serological Pipettes
Nunc Serological Pipettes offer precise liquid handling for reliable lab results. Designed for accuracy and ease of use, they feature clear graduations and a smooth draw. They are sterile and RNase/DNase-free, ensuring contamination-free experiments.
Overview
63.09 GBP valid until 2025-12-31
Use promo code "24071" to get your promotional price.
Additional Details:
Additional Details: Weight: 3.90600kg
Product Code. 16440901
Quantity Price
1 £ 138.0 / Pack of 200
In stock, ready to ship: 44
In stock, ready to ship: 959
Add to basket
  • Description and Specification
  • Alternative Products
  • Special Offers
Description and Specification
Specification
Accuracy ±1% at full volume
Aspirator Plug Yes
Autoclavable Not Autoclavable
Certifications/Compliance Rnase and DNase free, human DNA free, non-cytotoxic, non-pyrogenic
Description Serological Pipette, 5 year shelf life
Material Polystyrene
Sterility Sterile
For Use With (Application) Cell Culture
Graduated Yes
Quantity 50/Pk., 200/Cs.
Packaging Type Individual Paper/Plastic Wrap
Color Clear
Volume (Metric) 25 mL
Length (Metric) 276 mm
Type Serological Pipet
Disposable Single-use
Color Code Red
  • Wide range of volumes and packaging option
  • Color-coded pipettes and packaging for convenient volume selection
  • Individually packed pipettes in either paper/plastic or plastic peel for high sterility assurance
  • Reduce packaging waste with bulk packed options
Certified calibration of accuracy and precise graduations
  • Ensures dispensing accuracy +/-2% for 1 mL and 2 mL and +/-1% for 5 mL, 10 mL, 25 mL, and 50 mL at full volume
Clear, bold, bi-directional graduations
  • High contrast graduations for easy-to-read volumes
  • Reverse graduations aid in determining dispensing volume
Negative graduations
  • Increase pipette capacity and accommodate additional volumes
Filter plug
  • Prevent aerosol or liquid contamination of the pipetting device while further reducing sample-to-sample contamination risks
Product testing
  • Certificate of Quality
Additional information
  • Material: Ultra clear, 100% virgin polystyrene for maximum clarity
  • Contaminant free: RNase/DNase free, human DNA free, non-cytotoxic, non-pyrogenic
  • Sterility: e-beam irradiation to SAL of 10-6