Use promo code "24124" to get your promotional price.
Applied Biosystems™ MicroAmp™ TriFlex 96-Well PCR Reaction Plate
The Applied Biosystems MicroAmp TriFlex 96-Well PCR Reaction Plate is a versatile polypropylene PCR plate designed for use with Applied Biosystems standard (0.2 mL) 96-well and ProFlex 3x32-well thermal blocks.
100.49 GBP valid until 2025-08-31
Use promo code "24124" to get your promotional price.
Use promo code "24124" to get your promotional price.
Additional Details:
Weight:
0.57500kg
Specification
Barcode | No Barcode |
---|---|
Certifications/Compliance | Certified DNA/RNase Free |
Content And Storage |
Store at room temperature. |
Packaging | 20 Plates |
Product Line | MicroAmp |
Color | Optical |
Product Type | Microplate |
For Use With (Equipment) | MiniAmp™ Plus Thermal Cycler, MiniAmp™ Thermal Cycler, SimpliAmp™ Thermal Cycler, Veriti™ Thermal CyclerGeneAmp 9700, Veriti™ Thermal Cycler, ProFlex™ PCR System, 2720 Thermal Cycler, Veriti™ Dx Thermal Cycle |
Skirt Style | Semi |
Material | Polypropylene |
Volume (Metric) Well | 0.3 mL |
No. of Wells | 96 |
Volume (Metric) Working | 0.2 mL |
Quantity | 20 plates |
Shipping Condition | Room Temperature |