Thermo Scientific™ Multifuge X4 and X4R Pro Centrifuges
High footprint-to-capacity ratio with a Pro series touch screen for faster processing and programming, ideal for multi-use applications requiring more rotor choices, speeds and programming capacity.
Additional Details:
Weight:
120.00000kg
Specification
Certifications/Compliance | EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15 |
---|---|
Controller Type | Microprocessor |
Capacity | 4 x 1000 mL BioBottles, 196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 4 x 6 Microplates with TX-1000 Rotor |
Max. RCF | 25,830 x g Fixed Angle with Microliter 30 x 2 rotor, 7,197 x g Swing-Out with H-Flex HS4 rotor |
Product Line | Multifuge |
Refrigerated | Yes |
Display | Full Color Touch Screen Interface |
Drive System | Direct, Brushless Induction Low Profile |
For Use With (Equipment) | Purchase of Thermo Scientific rotor and accessories required for operation |
Green Features | Energy efficient, less waste |
Includes | Printed English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity |
Material | Front, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber |
Memory | Stores up to 100 events and programs |
Model | Multifuge X4R Pro-MD, IVD Certified |
Noise Level | <64 dB(A) Rotor F15-8x50c; 14500 RPM |
Profile (Acceleration/Braking) | Choices of 9 Accel/10 Decel Rates |
Program Storage | Up to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options |
Rotor | 18 Rotor options available |
Rotor Included | No |
Run Time | Up to 99 hr. 59 min. 59 sec. |
Standards | ISO 14971, ISO 13485, ISO 9001 |
Temperature Range | -10°C to +40°C, ±2°C |
Timer | Option to count up or down and to set “At start” or “At speed” |
Type | Benchtop Centrifuge |
Ventilated | No |
Warranty | 5 Year Limited |
Dimensions (L x W x H) Exterior | 690 x 746 x 361 mm |
Length (English) Exterior | 27.2 in. |
Length (Metric) Exterior | 690 mm |
Width (English) Exterior | 29.4 in. |
Width (Metric) Exterior | 746 mm |
Height (English) Exterior | 14.2 in. |
Height (English) Exterior Lid Open | 33.9 in. |
Height (Metric) Exterior | 361 mm |
Height (Metric) Exterior Lid Open | 861 mm |
Depth (English) | 27.2 in. |
Depth (Metric) | 690 mm |
Net Weight | 120 kg |
Electrical Requirements | 220-240 V 50 Hz / 230 V 60 Hz |
Voltage | 220-240 V |
Wattage | 1950 w |
Frequency | 50/60 Hz |
Laboratory/Cleanroom Use | Cleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1 |
-
Equipment End of Year Sale
Don't miss our end-of-year sale: save up to 50% on premium lab equipment and instruments.
Buy Now