166.83 GBP valid until 2025-08-24
Use promo code "24117" to get your promotional price.
Use promo code "24117" to get your promotional price.
| Binding Capacity | Low binding |
|---|---|
| Certifications/Compliance | Non-pyrogenic per USP Standard Lot-to-Lot |
| Disposable | Single-use |
| Material | PES |
| Material (Membrane) | PES |
| Pore Size | 0.20 μm |
| Sterility | Sterile |
| For Use With (Application) | Sterile Filtration of media and serum |
| Type | Filter Unit |
| Volume (Metric) | 1000 mL |
| Diameter (Metric) Membrane | 90 mm |