Honeywell Riedel-de Haën Dichloromethane, For pesticide residue analysis, Honeywell Riedel-de Haën™
Quantity | 4 x 2.5 L |
---|
Chemical Identifiers
CAS | 75-09-2 |
---|---|
Molecular Formula | CH2Cl2 |
Molecular Weight (g/mol) | 84.93 |
MDL Number | MFCD00000881 |
InChI Key | YMWUJEATGCHHMB-UHFFFAOYSA-N |
Synonym | methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm |
PubChem CID | 6344 |
ChEBI | CHEBI:15767 |
IUPAC Name | dichloromethane |
SMILES | ClCCl |
Specification
Chemical Name or Material | Dichloromethane |
---|---|
Melting Point | -97°C (literature) |
Name Note | For pesticide residue analysis |
CAS | 75-09-2 |
CAS Min % | ≥99.0000% |
CAS Max % | ≥99.0000% |
Density | 1.325 g/mL at 25 °C(lit.) |
Boiling Point | 39°C to 41°C |
Quantity | 4 x 2.5 L |
Packaging | Glass Bottle |
Molecular Formula | CH2Cl2 |
Linear Formula | CH2Cl2 |
MDL Number | MFCD00000881 |
UN Number | UN1593 |
Beilstein | 1730800 |
Synonym | methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm |
Synonym | methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm |
Synonym | methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm |
Synonym | methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm |
Synonym | methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm |
Synonym | methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm |
Synonym | methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm |
Synonym | methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm |
Synonym | methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm |
Synonym | methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm |
InChI Key | YMWUJEATGCHHMB-UHFFFAOYSA-N |
SMILES | ClCCl |
IUPAC Name | dichloromethane |
Molecular Weight (g/mol) | 84.93 |
PubChem CID | 6344 |
ChEBI | CHEBI:15767 |
Formula Weight | 84.93 |
Percent Purity | >99.8% |
Grade | Pesticide |
Technical documents
[Dichloromethane]
[Warning]
[{code=SKIN IRRIT. 2, sequence=1, description=Skin corrosion/irritation Category 2}, {code=EYE DAM. 2, sequence=2, description=Serious eye damage/eye irritation Category 2}, {code=CARC. 2, sequence=3, description=Carcinogenicity Category 2}, {code=STOT SE 3, sequence=4, description=Specific target organ toxicity Category 3}]
[{code=H315, sequence=1, description=Causes skin irritation.}, {code=H319, sequence=2, description=Causes serious eye irritation.}, {code=H336, sequence=3, description=May cause drowsiness or dizziness.}, {code=H351, sequence=4, description=Suspected of causing cancer.}]
[{code=P260, sequence=51, description=Do not breathe dust/fume/gas/mist/vapours/spray.}, {code=P280, sequence=52, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P302+P352, sequence=53, description=IF ON SKIN: Wash with plenty of water/.}, {code=P304+P340, sequence=54, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P305+P351+P338, sequence=55, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P308+P313, sequence=56, description=IF exposed or concerned: Get medical advice/attention.}]
[{code=GHS07, sequence=1, description=EXCLAMATION MARK}, {code=GHS08, sequence=2, description=HEALTH HAZARD}]
Product Identifier
- Dichloromethane
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315- Causes skin irritation.
- H319- Causes serious eye irritation.
- H336- May cause drowsiness or dizziness.
- H351- Suspected of causing cancer.
Precautionary Statement
- P260- Do not breathe dust/fume/gas/mist/vapours/spray.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352- IF ON SKIN: Wash with plenty of water/.
- P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313- IF exposed or concerned: Get medical advice/attention.