3M™ Solus™ 1000 Safety Glasses, Scotchgard™ Anti-Fog / Anti-Scratch Coating (K&N)
3M™ Solus™ Safety Glasses, feature a clear, strong polycarbonate lens in a slim, stylish, green and black-accented colour frame. Our safety glasses have 3M™ Scotchgard™ Anti-Fog / Anti-Scratch Coating, and a soft temple and nose bridge.
Overview
200.82 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Code: NEW
Additional Details:
Additional Details: Weight: 1.00000kg
Product Code. 15556565
Quantity Price
1 £ 352.0 / Pack of 20
Estimated Shipment
24-11-2025
Add to basket
  • Description and Specification
  • Alternative Products
Description and Specification
Specification
Certifications/Compliance ANSI Z87.1+ 2015, CSA Z94.3
Lens Coating Anti-fog
Lens Tint Amber
Product Type Safety Glasses
For Use With (Application) Construction, Food and Beverage Manufacturing, General Manufacturing, Metal Fabrication, Military Maintenance, Repair and Operation (MRO), Oil & Gas, Primary Metals, Transportation
Frame Color Blue, Black
Model S1103SGAF
Temple Style Removable
Lens Material Polycarbonate
Frame Material Polycarbonate
Frame Type Half Frame
  • Stylish and modern look slim frame with colour accented soft temple components
  • Strong polycarbonate lens for impact resistance
  • 3M™ Scotchgard™ Anti-Fog Coating offers superior anti-fog and anti-scratch properties
  • Lightweight with soft cushioning on nose bridge
  • Inner padding on temples for increased comfort and stability when worn
  • Meets K&N marking requirements of EN 166
  • Frame marking: 3M EN166 FT CE
Compliance: Meets ANSI Z87.1+ 2015 and CSA Z94.3 standards.
Alternative Products