Fisherbrand™ Mini Vortexer, Variable Speed Mixer 500-2800 rpm
Small, compact vortex mixer
Overview
107.15 GBP valid until 2025-07-31
Use promo code "24075" to get your promotional price.
Code: NEW
Additional Details:
Additional Details: Weight: 0.93780kg
Product Code. 15494960
Quantity Price
1 £ 268.0 / Each
In stock, ready to ship: 6
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
  • Special Offers
Description and Specification
Specification
Orbit 4.5 mm
Speed Range 500 to 2800 rpm
Plug Type EU, UK Plug
  • Adjustable speed from 500 to 2,800rpm
  • Orbit 4.5mm
  • Touch-function activates the motor immediately upon contact with a tube or vessel
  • The unique cup design accepts tubes and vessels of all sizes, including 15 and 50mL conical tubes
  • Power supply - supplied with both UK and EU adapters
  • Two year warranty
Alternative Products
Documentation Special Offers