Use promo code "24128" to get your promotional price.
200MG Rapamycin, 99+%
285.48 GBP valid until 2025-08-31
Use promo code "24128" to get your promotional price.
Use promo code "24128" to get your promotional price.
200MG Rapamycin, 99+%
Code:
NEW
Additional Details:
CAS Number:
53123-88-9
Weight:
0.00020kg
Technical documents
[Rapamycin]
[Danger]
[{code=H351, sequence=1, description=Suspected of causing cancer.}, {code=H361, sequence=2, description=Suspected of damaging fertility or the unborn child.}, {code=H372, sequence=3, description=Causes damage to organs.}]
[{code=P201, sequence=51, description=Obtain special instructions before use.}, {code=P280, sequence=52, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P308+P313, sequence=53, description=IF exposed or concerned: Get medical advice/attention.}]
[{code=MIXTURE LIST, sequence=1, description=Contains : Sirolimus}]
[{code=GHS08, sequence=1, description=HEALTH HAZARD}]
Product Identifier
- Rapamycin
Signal Word
- Danger
Hazard Statement
- H351- Suspected of causing cancer.
- H361- Suspected of damaging fertility or the unborn child.
- H372- Causes damage to organs.
Precautionary Statement
- P201- Obtain special instructions before use.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313- IF exposed or concerned: Get medical advice/attention.
Supplemental Information
- MIXTURE LIST- Contains : Sirolimus