Code:
NEW
Thermo Scientific™ Armadillo PCR Plate, 96-well, blue, semi-skirted, low profile, clear wells
Thermo Scientific Armadillo PCR, Blue Semi-Skirted, Low Profile, Clear Well plates are the ultimate in high performance PCR plastics. Combining the rigidity of a polycarbonate frame with thin walled polypropylene wells, the Armadillo plate provides superior thermal cycling performance under all conditions without warping.
Additional Details:
Weight:
0.59000kg
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specification
Barcode | No Barcode |
---|---|
Certifications/Compliance | Certified RNase/DNase-free |
Packaging | 25 Plate |
Product Line | Armadillo |
Color | Blue,Clear |
Color | Blue,Clear |
Product Type | PCR Microplate |
For Use With (Equipment) | Real-Time PCR Systems or Thermal Cyclers |
Skirt Style | Semi |
Material | Polycarbonate,Polypropylene |
Material | Polycarbonate,Polypropylene |
Volume (Metric) Well | 0.2 mL |
No. of Wells | 96 |
Volume (Metric) Working | 0.1 mL |
Quantity | 25 plates |
Reaction Speed | Low profile |