Use promo code "24079" to get your promotional price.
Invitrogen™ Bolt™ Bis-Tris Plus Mini Protein Gels, 10%, 1.0 mm, WedgeWell™ format
Similar to NuPAGE™ Bis-Tris gels, Bolt™ Bis-Tris Plus gels are designed to deliver consistent gel performance and provide a neutral pH environment that minimizes protein modifications.
110.20 GBP valid until 2025-12-31
Use promo code "24079" to get your promotional price.
Use promo code "24079" to get your promotional price.
Code:
NEW
Additional Details:
Weight:
1.07000kg
Requires:
Mini Gel Tank (Mfr. No. A25977)
Specification
For Use With (Equipment) | Mini Gel Tank,XCell SureLock Mini-Cell |
---|---|
For Use With (Equipment) | Mini Gel Tank,XCell SureLock Mini-Cell |
Gel Percentage | 10% |
Gel Size | Mini |
GelThickness | 1.0 mm |
Gel Type | Bis-Tris Plus |
Recommended Applications | Denaturing |
Sample Loading Volume | up to 30 μL |
Separation Range | 3.5 to 260 kDa |
Separation Type | Denaturing |
Wells | 17-well |
Mode of Separation | Molecular Weight |
Product Line | Bolt™, WedgeWell™ |
Quantity | 10 gels (1 box) |
Shipping Condition | Room Temperature |
Storage Requirements | Store at 4–25°C. Do not freeze. |
Length (Metric) | 8 cm |
Width (Metric) | 8 cm |
-
Protein Biology Essentials Promotion
Our top products empowering your research from start to finish
Buy Now