102.93 GBP valid until 2025-09-30
Use promo code "24076" to get your promotional price.
Use promo code "24076" to get your promotional price.
Detection Method | Fluorescence |
---|---|
For Use With (Equipment) | Qubit Fluorometer |
For Use With (Application) | microRNA Quantitation |
No. of Reactions | 100 Reactions |
Product Line | Qubit |
Product Type | microRNA Assay Kit |
Quantity | 100 assays |
Shipping Condition | Room Temperature |
Excitation/Emission | 498/518 |