Code:
Z3
Diba Omnifit™ T-Series Bottle Caps
Easy to use and versatile range of caps requires no fittings
Additional Details:
Weight:
0.30000kg
Specification
Color | Blue |
---|---|
Includes | 3 Ports, No Valve |
Material | PTFE |
No. per Case | 1 Cs. |
Sterility | Non-sterile |
Thread Size (Metric) | GL 38 |
Neck Finish | 38-430 |
Autoclavable | Not Autoclavable |
Disposable | Reusable |
Shape | Round |
Height (Metric) | 28.5 mm |
Product Type | Solvent Waste Cap |
For Use With (Equipment) | T series solvent bottle |
Height (English) | 1.122 in. |
Diameter (Metric) | 54 mm |