Waters Corporation XBridge Shield RP18 Column
XBridge BEH Shield RP18 columns complement the XBridge BEH C18 and C8 phases and provide a necessary and powerful tool for HPLC method development.
Additional Details:
Weight:
0.50000kg
Specification
Diameter (Metric) | 1 mm |
---|---|
Length (Metric) | 150 mm |
Max. Pressure | 6000 psi (415 Bar) |
Particle Shape | Spherical |
Particle Size | 3.5 μm |
Pore Size | 130 Å |
Quantity | 1/Ea. |
USP Type | L1 |
Endcapped | Yes |
Type | Column |