Use promo code "25339" to get your promotional price.
Acetonitrile, 99.6%, ACS reagent
Use promo code "25339" to get your promotional price.
| Quantity | 10 L |
|---|
Chemical Identifiers
| CAS | 75-05-8 |
|---|---|
| Molecular Formula | C2H3N |
| Molecular Weight (g/mol) | 41.053 |
| MDL Number | MFCD00001878 |
| InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
| Synonym | methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitrile,cyanure de methyl,methylkyanid,mecn |
| PubChem CID | 6342 |
| ChEBI | CHEBI:38472 |
| IUPAC Name | acetonitrile |
| SMILES | CC#N |
Specification
| Chemical Name or Material | Acetonitrile |
|---|---|
| Melting Point | -46.0°C |
| Name Note | For analysis ACS |
| CAS | 75-05-8 |
| CAS Min % | 99.5 |
| CAS Max % | 100.0 |
| Decomposition Information | >500°C |
| Density | 0.7810g/mL |
| Quantity | 10 L |
| Flash Point | 2°C |
| Physical Form | Liquid |
| Refractive Index | 1.3430 to 1.3450 |
| Residue after Evaporation | 0.005% max. |
| Assay Percent Range | 99.6% |
| Specific Gravity | 0.781 |
| Titratable Acid | 0.008 meq/g max. |
| Molecular Formula | C2H3N |
| Titratable Base | 0.0006 meq/g max. |
| Linear Formula | CH3CN |
| Vapor Pressure | Vapor Pressure: 97mbar at 20°C |
| MDL Number | MFCD00001878 |
| Viscosity | 0.36 mPa.s (20°C) |
| Water | 0.3% max. (K.F.) |
| Beilstein | 02, 183 |
| Fieser | 02,13; 12,450; 13,156; 15,1 |
| Merck Index | 15, 69 |
| Synonym | methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitrile,cyanure de methyl,methylkyanid,mecn |
| Synonym | methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitrile,cyanure de methyl,methylkyanid,mecn |
| Synonym | methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitrile,cyanure de methyl,methylkyanid,mecn |
| Synonym | methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitrile,cyanure de methyl,methylkyanid,mecn |
| Synonym | methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitrile,cyanure de methyl,methylkyanid,mecn |
| Synonym | methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitrile,cyanure de methyl,methylkyanid,mecn |
| Synonym | methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitrile,cyanure de methyl,methylkyanid,mecn |
| Synonym | methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitrile,cyanure de methyl,methylkyanid,mecn |
| Synonym | methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitrile,cyanure de methyl,methylkyanid,mecn |
| Synonym | methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitrile,cyanure de methyl,methylkyanid,mecn |
| Boiling Point | 81.0°C to 82.0°C |
| Solubility Information | Solubility in water: miscible. Other solubilities: soluble in alcohol, ethyl acetate, acetone, ether |
| InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
| SMILES | CC#N |
| IUPAC Name | acetonitrile |
| Molecular Weight (g/mol) | 41.053 |
| PubChem CID | 6342 |
| ChEBI | CHEBI:38472 |
| Formula Weight | 41.05 |
| Percent Purity | 99.6% |
| Grade | ACS |
Technical documents
[Acetonitrile]
[Danger]
[{code=ACUTE TOX. 4, sequence=1, description=Acute toxicity Category 4}, {code=EYE DAM. 2, sequence=2, description=Serious eye damage/eye irritation Category 2}, {code=FLAM. LIQ. 2, sequence=3, description=Flammable liquid Category 2}]
[{code=H319, sequence=1, description=Causes serious eye irritation.}, {code=H302, sequence=2, description=Harmful if swallowed.}, {code=H312, sequence=3, description=Harmful in contact with skin.}, {code=H332, sequence=4, description=Harmful if inhaled.}, {code=H225, sequence=5, description=Highly flammable liquid and vapour.}]
[{code=P210, sequence=50, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P280, sequence=51, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P301+P312, sequence=52, description=IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.}, {code=P302+P352, sequence=53, description=IF ON SKIN: Wash with plenty of water/.}, {code=P304+P340, sequence=54, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P305+P351+P338, sequence=55, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS07, sequence=2, description=EXCLAMATION MARK}]
Product Identifier
- Acetonitrile
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
Hazard Statement
- H319- Causes serious eye irritation.
- H302- Harmful if swallowed.
- H312- Harmful in contact with skin.
- H332- Harmful if inhaled.
- H225- Highly flammable liquid and vapour.
Precautionary Statement
- P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312- IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352- IF ON SKIN: Wash with plenty of water/.
- P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.