Thermo Scientific™ Finnpipette™ F3 Variable Volume Single Channel Pipettes
Support every day pipetting in comfort and with ideal function using these manual variable volume pipettes.
Overview
191.83 GBP valid until 2025-07-31
Use promo code "24075" to get your promotional price.
Code: 04
Additional Details:
Additional Details: Weight: 0.29000kg
Product Code. 11815231
Quantity Price
1 £ 285.0 / Each
In stock, ready to ship: 79
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
  • Special Offers
Description and Specification
Specification
Accuracy ±3.5 to 1.0%
Compatible Tips Finntip 250, Filter 10, Flex 200, Flex Filter 30, 200 Ext, Wide 250
Description Finnpipette F3 1-10μL
Color Yellow
Model F3
Number of Channels 1
Product Line Finnpipette™ F3
Type Variable Volume Pipette
Volume (Metric) 1 to 10 μL
Increments 0.02μL
Precision 3.0 to 0.8%
Color Code Yellow
Ergonomic Comfort
  • Wide color-coded supportive finger rest for identification support and ergonomics
  • Designed handle softly contours the shape of your hand
  • Large Ergovisio Display for volume recognition
Ideal Function
  • Durable handle material
  • Double-action pipetting button; rotating upper part prevents accidental volume alteration
  • Advanced volume gearing mechanism for enhanced precision and accuracy
  • Super blow-out for efficient delivery of micro-size drops in 50ul models and below
Compatible with Finntip pipette tips, and other tip models
Alternative Products
Documentation Special Offers