DWK Life Sciences Wheaton™ Polypropylene Caps for Wheaton Dropping Bottles - Yellow
Screw cap made of radiation grade polypropylene
Additional Details:
Weight:
0.47630kg
Specification
Color | Yellow |
---|---|
Material | Polypropylene |
No. per Case | 100 Cs. |
Neck Finish | 13-425 |
Certifications/Compliance | ISO 9001 |
Product Type | Polypropylene Cap |
For Use With (Equipment) | Plastic dropping bottle |