Code:
A0
Cytiva Whatman™ Grade 5V Fluted Fine Particle Filter Paper, 185mm circle
GE’s Whatman Grade 5V fine particle filter retains particles as small as 2.5 μm for qualitative analysis. This cellulose filter paper is fluted for ease-of-use.
Additional Details:
Weight:
0.28000kg
Specification
Description | Grade 5V Fluted Fine Particle Filter Paper, 185 mm circle (100 pcs) |
---|---|
Format | Pre-pleated Circle |
Material | Cellulose |
Pore Size | 2.5 μm |
Filtration Speed | 1420 sec. |
Flow Rate | Slow |
For Use With (Application) | GE’s Whatman Grade 5V fine particle filter retains particles as small as 2.5 μm for qualitative analysis. This cellulose filter paper is fluted for ease-of-use. |
Grade | Grade 5V |
Porosity | Fine |
Quantity | 100 Pk. |
Thickness (Metric) | 200 μm |
Diameter (Metric) | 185 mm |