| Chemical Name or Material |
Ethyl acetate
|
| Melting Point |
-84°C
|
| CAS |
141-78-6
|
| Color |
Colorless
|
| Density |
0.902 g/mL
|
| Boiling Point |
77°C
|
| Quantity |
1 L
|
| Flash Point |
-3°C (26°F)
|
| Odor |
Sweet
|
| Physical Form |
Liquid
|
| Refractive Index |
1.372
|
| Molecular Formula |
C4H8O2
|
| Linear Formula |
CH3CO2CH2CH3
|
| MDL Number |
MFCD00009171
|
| UN Number |
UN1173
|
| Beilstein |
506104
|
| Merck Index |
14,3757
|
| Synonym |
ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester
|
| Synonym |
ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester
|
| Synonym |
ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester
|
| Synonym |
ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester
|
| Synonym |
ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester
|
| Synonym |
ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester
|
| Synonym |
ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester
|
| Synonym |
ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester
|
| Synonym |
ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester
|
| Synonym |
ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester
|
| Solubility Information |
Miscible with ethanol,acetone,diethyl ether and benzene.
|
| InChI Key |
XEKOWRVHYACXOJ-UHFFFAOYSA-N
|
| SMILES |
CCOC(=O)C
|
| IUPAC Name |
ethyl acetate
|
| Molecular Weight (g/mol) |
88.106
|
| PubChem CID |
8857
|
| ChEBI |
CHEBI:27750
|
| Formula Weight |
88.11
|
| Percent Purity |
≥99.5%
|
| Grade |
ACS
|