184.39 GBP valid until 2025-08-31
Use promo code "24124" to get your promotional price.
Use promo code "24124" to get your promotional price.
Barcode | No Barcode |
---|---|
Certifications/Compliance | RNase-, DNase-free |
Format | Standard |
Lid | Without Lid |
Array | 8 x 12 |
Color | White |
For Use With (Equipment) | Roche Light Cycler 480 |
Material | Polypropylene |
No. of Wells | 96 |
Sterility | Non-sterile |
Well Shape | Round |
For Use With (Application) | PCR thermal transfer, RT-PCR applications |
Includes | Printed alphanumeric grid |
No. per Case | 50/Cs. |
No. per Pack | 10/Pk. |