Use promo code "25339" to get your promotional price.
Thermo Scientific™ Nalgene™ Colored Polypropylene Closures with 38-430 Finish
Ensure leakproof performance with these colored, polypropylene closures with 38-430 finish. Closures are one-piece and linerless with integrally molded seal ring.
89.30 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Use promo code "25339" to get your promotional price.
Code:
A0
Additional Details:
Weight:
1.00000kg
Specification
Color | Green |
---|---|
Description | Nalgene Colored Closures |
Material | Polypropylene |
No. per Case | 250 |
No. per Pack | 250 |
Packaging | Bulk |
Thread Size (Metric) | 38-430 |
Liner Material | Unlined |
Certifications/Compliance | Manufactured in compliance with ISO 13485:2016 quality requirements. Closures meet FDA CFR21 177.1520 for food and beverage use, USP Class VI. Meet the requirements of light-resistant containers per USP latest edition when used with opaque bottles. |
Mouth | Narrow |
Shape | Round |
Height (Metric) | 27 mm |
Product Type | Screw Cap |
For Use With (Equipment) | Nalgene Narrow-Mouth 1000mL Bottle |
Height (English) | 1.06 in. |
Diameter (Metric) | 41 mm (Outer) |