355.91 GBP valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Use promo code "24156" to get your promotional price.
Capacity (English) | 8.45 to 12.84 oz. |
---|---|
Certifications/Compliance | USP Class VI |
Closure Color | Orange |
Diameter (Metric) Closure | 33mm |
Capacity (Metric) | 250 to 380 mL |
Material | Polystyrene |
Product Type | Culture Chamber |
Graduated | Yes |
Culture Area | 1272 cm² |
Surface Treatment | Tissue Culture-treated |
Quantity | Two chambers |
Sterilization Method | Gamma Irradiated |