Code:
50
Macherey-Nagel™ Optima™ 17 GC Capillary Column
Additional Details:
Weight:
0.01000kg
Specification
| For Use With (Application) | Steroids Analysis,Pesticide Analysis,Drug Analysis |
|---|---|
| For Use With (Application) | Steroids Analysis,Pesticide Analysis,Drug Analysis |
| For Use With (Application) | Steroids Analysis,Pesticide Analysis,Drug Analysis |
| Film Thickness | 0.25 μm |
| Length (Metric) | 60 m |
| Quantity | 1 Each |
| Diameter (Metric) | 0.5 mm |
| USP Type | G1, G2, G38 |
| Stationary Phase | Optima™ 17 |
| Type | GC Capillary Column |
| Diameter (Metric) Inner | 0.32 mm |
| Temperature Range (Metric) | 320°C to 340°C |