179.43 GBP valid until 2025-08-31
Use promo code "24124" to get your promotional price.
Use promo code "24124" to get your promotional price.
Certifications/Compliance | ISO 9001:2000, FDA, CFR |
---|---|
Closure Color | Blue |
Diameter (English) Outside | 1.2 in. |
Color | Clear |
Capacity (Metric) | 50 mL |
Material | Polypropylene |
For Use With | For DNA isolation, purification and precipitation of nucleic Acids, cell centrifugation applications |
No. per Case | 500 |
Packaging | Rack |
Sterility | Sterile |
Shape | Conical |
Length (Metric) | 115 mm |
Capacity (English) | 1.69 oz. |
Bottom Shape | Conical |
Writing Area | White Marking Spot |
Diameter (Metric) | 30 mm |
Product Type | Centrifuge Tube |
Max. RCF | 16,000 x g |