Code:
NEW
Corning™ 384-Well, Cell Culture-Treated, Flat-Bottom, Optical Imaging Microplate
Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Lid
Additional Details:
Weight:
10.79549kg
Specification
Format | Standard |
---|---|
Lid | With Lid |
Culture Area | 0.06 cm2 |
Array | 16 x 24 |
Color | Black,Clear Bottom |
Color | Black,Clear Bottom |
Material | Polystyrene |
No. of Wells | 384 |
Volume (Metric) Working | 20 to 80 μL |
Sterility | Sterile |
Surface Treatment | Cell Culture |
Total Volume (Metric) Well | 112 μL |
Well Shape | Flat Bottom (F-well) |
Includes | With generic barcode |
No. per Case | 100/Cs. |
No. per Pack | 20/Pk. |
Quantity | 100/Cs. |
Well Color | Clear |